Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPA1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11055290C
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
OPA1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting | |
O60313 | |
OPA1 | |
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat and chicken. Predicted to react with primate based on 100% sequence homology. Immunogen sequence has 94% homology to Zebrafish. | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
Rabbit | |
111 kDa | |
0.1 mL | |
Apoptosis, Mitochondrial Fusion Proteins, Neurodegeneration, Neuroscience, Sensory Systems, Vision | |
4976 | |
Human, Mouse, Rat, Pig, Chicken | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction