Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPA1 Monoclonal Antibody (1E8-1D9)
GREENER_CHOICE

Catalog No. PIMA516149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516149 Supplier Invitrogen™ Supplier No. MA516149
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot, multiple protein isoforms can be seen at approximately 90, 80 and 65 kDa.

OPA1 is a dynamin-related GTPase that is critical for the maintenance of mitochondrial morphology and mtDNA. The most commonly associated phenotype with OPA1 mutations is heterozygous optic atrophy, a heterozygous dominant trait that causes reduced visual clarity and sometimes blindness. The disease usually begins in childhood and increases in severity throughout the life of affected individuals. Usually, this phenotype is attributed to the degeneration of optic nerve fibers. Interestingly, the same type of nerve degeneration seems to be partially causative of certain schizophrenia characteristics. OPA1 dysfunction also seems to be implicated in this case; mitochondrial networks associated with critical nerves seem to link schizophrenia and OPA1. The dysfunction is associated with issues with apoptosis and normal cellular metabolic regulation, all regulated through OPA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1E8-1D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene OPA1
Gene Accession No. O60313, P58281, Q2TA68
Gene Alias 1200011N24Rik; AI225888; AI847218; BERHS; Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; EMBL:AAI11072.1}; hypothetical protein; KIAA0567; Large GTP-binding protein; largeG; lilr3; MGM1; mitochondrial dynamin-like GTPase; mitochondrial OPA1; mKIAA0567; MTDPS14; NPG; NTG; OPA1; opa1 {ECO:0000312; OPA1 mitochondrial dynamin like GTPase; OPA1, mitochondrial dynamin like GTPase; optic atrophy 1; optic atrophy 1 (autosomal dominant); optic atrophy 1 homolog; optic atrophy 1-like protein; optic atrophy protein 1; Optic atrophy protein 1 homolog; RCJMB04_1m16; RN protein
Gene Symbols OPA1
Host Species Mouse
Immunogen Human OPA1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100751873, 171116, 4976, 524142, 74143
Target Species Human, Mouse, Rat, Bovine, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.