Learn More
OPA3 Mouse anti-Human, Clone: OTI3G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809426

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The mouse ortholog of this protein co-purifies with the mitochondrial inner membrane. Mutations in this gene have been shown to result in 3-methylglutaconic aciduria type III and autosomal domit optic atrophy and cataract. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
OPA3 | |
Monoclonal | |
Unconjugated | |
OPA3 | |
MGA3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80207 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H6K4 | |
OPA3 | |
Full length human recombit protein of human OPA3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.