Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545032 Supplier Thermo Scientific Supplier No. PA545032
Add to Cart
Add to Cart

OPA3 Polyclonal Antibody

OPA3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OPA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OPA3
Gene Accession No. Q9H6K4
Gene Alias D630048P19Rik; FLJ22187; FLJ25932; Gm1425; Gm472; MGA3; MGC75494; OPA3; OPA3 outer mitochondrial membrane lipid metabolism regulator; optic atrophy 3; optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia); Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'); optic atrophy 3 protein; Optic atrophy 3 protein homolog; outer mitochondrial membrane lipid metabolism regulator OPA3
Gene Symbols OPA3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human OPA3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80207
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.