Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPAL1 Antibody (OPAL1-01), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262210H
Description
OPAL1 Monoclonal specifically detects OPAL1 in Human samples. It is validated for Western Blot.Specifications
OPAL1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
WBP1L | |
Recombinant fragment of human OPAL1 (amino acids 152-342) | |
0.1 mL | |
54838 | |
Human | |
IgG2a |
Western Blot | |
OPAL1-01 | |
Western Blot | |
chromosome 10 open reading frame 26, FLJ20154, FLJ20367, MGC125648, OPA1L, OPAL1, outcome predictor in acute leukemia 1 | |
Mouse | |
Protein A purified | |
Primary | |
The antibody HCG-61 reacts with beta-subunit of human chorionic gonadotropin, a 40 kDa hormone. hCG belongs to the glycoprotein hormone family together with lutropin (LH), follitropin (FSH) and thyrotropin (TSH). hCG is synthesised by trophoblastic cells of the placenta during pregnancy and stimulates the growth of corpus luteum. Following cross-reactivity (%) was determined by classic double-antibody RIA using unlabelled hormones: beta-hCG (77), alpha-hCG (1.3), hLH (0.86), hFSH (< 0.5), hTSH (< 0.5). | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction