Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPCML Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547392
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547392 Supplier Invitrogen™ Supplier No. PA547392
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) KILON and rhNeurotrimin is observed and less than 1% cross-reactivity with rhALCAM, rhBCAM, rhEpCAM and rhNCAM is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Binds opioids in the presence of acidic lipids; probably involved in cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPCML
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene OPCML
Gene Accession No. Q14982
Gene Alias 2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like
Gene Symbols OPCML
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human OBCAM/OPCML Gly28-Asn322.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4978
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.