Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPCML Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550247 Supplier Invitrogen™ Supplier No. PA550247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total OBCAM / OPCML protein.

Binds opioids in the presence of acidic lipids; probably involved in cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPCML
Applications Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene OPCML
Gene Accession No. Q14982
Gene Alias 2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like
Gene Symbols OPCML
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human opioid binding protein/cell adhesion molecule-like.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330908, 4978
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.