Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPCML Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103152 Supplier Thermo Scientific Supplier No. PA5103152
Add to Cart
Add to Cart

OPCML Polyclonal Antibody

OPCML Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OPCML
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OPCML
Gene Accession No. P32736, Q14982
Gene Alias 2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like
Gene Symbols OPCML
Host Species Rabbit
Immunogen A synthesized peptide derived from human OPCML(Accession Q14982), corresponding to amino acid residues S279-A329.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116597, 330908, 4978
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.