Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Open Air Rockers

Ideal for cell culture work and are designed to be used in a variety of environmental conditions

$1688.00 - $1695.00


Products 2
Catalog Number Plug Type Certifications/Compliance Price Quantity & Availability  
Catalog Number Plug Type Certifications/Compliance Price Quantity & Availability  
88-861-025
View Documents
US Plug cCSAus
Each for $1,688.00
Only null left
Edge
Add to Cart
 
88-861-026
View Documents
EU, UK, CN Plug CE
Each for $1,695.00
Only null left
Add to Cart
 
Description

Description

  • Low profile design: Takes up less space and easily fits into most fume hoods and incubators. Plastic base offers durability.
  • Microprocessor control: The microprocessor control provides electronic tilt angle adjustment which allows user to easily adjust rocking angle from 1 to 15° while unit is operating. Precise speed control provides smooth, low-speed rocking motion down to 1 rpm.
  • Memory: Display will show last user settings, even after power has been turned off. 
  • Continue to run by last set speed and the remaining time when the power recovered.
  • LED display: Touch pad controls with easy-to-read, LED displays for speed, tilt angle, and time . Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been shut off.
  • Applications: Staining and destaining gels, hybridization procedures, hematology and blotting technique

Warranty and Services

2 years

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.