Learn More
MilliporeSigma™ Supelco™ Open Top Screw Caps
Catalog No. p-7204507 Shop All MilliporeSigma Supelco Products
Click to view available options
Closure Size:
13-425
15-425
Closure type screw cap with cut-out top to prevent contamination
  • Autoclavable
  • Easy sterilization
  • Used as a closure type for the containers used to contain substances in order to prevent contamination
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic
Septum Without septum
No. per Pack 200
Closure Size 15-425
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.