Learn More
OPG Rabbit anti-Human, Polyclonal, Bioss
SDP
Rabbit Polyclonal Antibody
Supplier:  Bioss BS0431R
Encompass_Preferred
Catalog No. 50-198-0270
Only null left
Add to Cart
Description
Description
Specifications
Specifications
OPG
Polyclonal
Unconjugated
Rabbit
Protein A
RUO
Human
Antibody
IgG
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
KLH conjugated synthetic peptide derived from mouse OPG.
100 μL
Primary
-20°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.