Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPHN1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299305MFV500
Description
OPHN1 Polyclonal antibody specifically detects OPHN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
OPHN1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ARHGAP41, mental retardation, X-linked 60, MRX60, oligophrenin 1, oligophrenin-1, oligophrenin-1, Rho-GTPase activating protein, OPN1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OPHN1. | |
0.1 mL | |
Neuroscience | |
4983 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction