Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPHN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | OPHN1 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
OPHN1 Polyclonal antibody specifically detects OPHN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
OPHN1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS (pH 7.0) | |
4983 | |
IgG | |
Antigen and protein A Affinity-purified |
Immunohistochemistry-Paraffin 1:200-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ARHGAP41, mental retardation, X-linked 60, MRX60, oligophrenin 1, oligophrenin-1, oligophrenin-1, Rho-GTPase activating protein, OPN1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OPHN1. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title