Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OPHN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598030
Description
OPHN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Stimulates GTP hydrolysis of members of the Rho family. Its action on RHOA activity and signaling is implicated in growth and stabilization of dendritic spines, and therefore in synaptic function. Critical for the stabilization of AMPA receptors at postsynaptic sites. Critical for the regulation of synaptic vesicle endocytosis at presynaptic terminals. Required for the localization of NR1D1 to dendrites, can suppress its repressor activity and protect it from proteasomal degradation.
Specifications
OPHN1 | |
Polyclonal | |
Unconjugated | |
OPHN1 | |
ARHGAP41; C130037N19Rik; mental retardation, X-linked 60; MRX60; oligophrenin 1; oligophrenin-1; oligophrenin-1, Rho-GTPase activating protein; Ophn1; Opn1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4983 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60890 | |
OPHN1 | |
Recombinant Human Oligophrenin-1 protein (1-270AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction