Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Opicinumab Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542069
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Opicinumab is under investigation in clinical trial NCT02641041 (Safety, Tolerability, and Pharmacokinetics of Single and Multiple Doses of BIIB033 in Healthy Japanese Participants).
Specifications
Opicinumab | |
Recombinant Monoclonal | |
Unconjugated | |
BIIB-033 | |
Human LINGO1/LRRN6A/LERN1. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
3.65 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A/G | |
RUO | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction