Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OPLAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555346
Description
Immunogen sequence: RELGFTHVSL SSEAMPMVRI VPRGHTACAD AYLTPAIQRY VQGFCRGFQG QLKDVQVLFM RSDGGLAPMD TFSGSSAVLS GPAGGVVGYS ATTY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
OPLAH catalyzes the cleavage of 5-oxo-L-proline to form L-glutamate coupled to the hydrolysis of ATP to ADP and inorganic phosphate. ATP + 5-oxo-L-proline + 2 H2O = ADP + phosphate + L-glutamate.
Specifications
| OPLAH | |
| Polyclonal | |
| Unconjugated | |
| OPLAH | |
| 1700010G02Rik; 5-OPase; 5-oxo-L-prolinase; 5-oxoprolinase; 5-oxoprolinase (ATP-hydrolysing); OPLA; Oplah; OPLAHD; pyroglutamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14841 | |
| OPLAH | |
| Recombinant protein corresponding to Human OPLAH. Recombinant protein control fragment (Product #RP-93927). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction