Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OPLAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111354
Description
Immunogen sequence: QANAELAVRD MLRAFGTSRQ ARGLPLEVSS EDHMDDGSPI RLRVQISLSQ GSAVFDFSGT GPEVFGNLNA.
OPLAH catalyzes the cleavage of 5-oxo-L-proline to form L-glutamate coupled to the hydrolysis of ATP to ADP and inorganic phosphate. ATP + 5-oxo-L-proline + 2 H2O = ADP + phosphate + L-glutamate.
Specifications
| OPLAH | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14841 | |
| OPLAH | |
| Recombinant Protein corresonding to Human OPLAH. Recombinant protein control fragment (Product #RP-106396). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| OPLAH | |
| 1700010G02Rik; 5-OPase; 5-oxo-L-prolinase; 5-oxoprolinase; 5-oxoprolinase (ATP-hydrolysing); OPLA; Oplah; OPLAHD; pyroglutamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction