Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPN1SW1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572216
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572216 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572216 Supplier Invitrogen™ Supplier No. PA572216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OPN1SW1 Polyclonal Antibody for Western Blot

Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPN1SW1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene opn1sw1
Gene Accession No. Q9W6A9
Gene Alias blue cone opsin; opn1sw1; opn1sw2; opsin 1 (cone pigments), short-wave-sensitive 1; opsin 1 (cone pigments), short-wave-sensitive, 2; opsin SWS-1; Opsin-1, short-wave-sensitive 1; seven transmembrane light receptor; short-wavelength-sensitive-1 cone opsin; SWS1; Ultraviolet cone photoreceptor pigment; ultraviolet-sensitive opsin; uvops; zfuv
Gene Symbols opn1sw1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 15-47 amino acids of DANRE opn1sw1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30582
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.