Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPN1SW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572677
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572677 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572677 Supplier Invitrogen™ Supplier No. PA572677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OPN1SW2 Polyclonal Antibody for Western Blot

Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPN1SW2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene opn1sw2
Gene Accession No. Q9W6A8
Gene Alias blue cone opsin; blue cone photoreceptor pigment; Blue-sensitive opsin; bluops; opn1sw1; opn1sw2; opsin 1 (cone pigments), short-wave-sensitive 2; opsin SWS-2; Opsin-1, short-wave-sensitive 2; seven transmembrane light receptor; short-wavelength-sensitive-2 cone opsin; SI:zK13A21.5; SWS2; zfblue
Gene Symbols opn1sw2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 242-276 amino acids of DANRE opn1sw2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30435
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.