Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPN4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572273
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572273 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572273 Supplier Invitrogen™ Supplier No. PA572273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OPN4L Polyclonal Antibody for Western Blot

Photoreceptor implicated in non-image-forming responses to light.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPN4L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene opn4.1
Gene Accession No. Q1JPS6
Gene Alias mammalian-like melanopsin-2; melanopsin; melanopsin opn4m2; melanopsin-like; opn4; opn4.1; opn4c; opn4l; opn4m2; opsin; opsin 4 (melanopsin); opsin 4, like; opsin 4.1; Opsin-4-like
Gene Symbols opn4.1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 255-288 amino acids from the Central region of DANRE opn4l.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352918
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.