Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121199 Supplier Invitrogen™ Supplier No. PA5121199
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: LHLCIALGYA NSSLNPVLYA FLDENFKRCF RQLCRKPCGR PDPSSFSRAR EATARERVTA CTPSDGPGGG AAA.

OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPRD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OPRD1
Gene Accession No. P32300, P33533, P41143
Gene Alias delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a
Gene Symbols OPRD1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-372 of human OPRD1 (NP_0009023).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18386, 24613, 4985
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.