Learn More
Invitrogen™ OPRD1 Polyclonal Antibody

Description
PA1-4558 detects delta Opioid Receptor from human, mouse, and rat samples. PA1-4558 has been successfully used in Western blotting applications. This antibody will detect a dominant band of ~48 kDa representing the monomer form of DOR-1, and may also detect dimmers or trimers as heterodimerization and homodimerization is common The PA1-4558 immunogen is a synthetic peptide corresponding to residues 3-17 L(3) V P S A R A E L Q S S P L V(17) of mouse and rat DOR-1.
Specifications
Specifications
| Antigen | OPRD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | OPRD1 |
| Gene Accession No. | P32300, P33533, P41143 |
| Gene Alias | delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a |
| Gene Symbols | OPRD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.