Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ oprI Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144555
Description
oprI Polyclonal Antibody for Western Blot, ELISA
OprI is a protein that is found in the outer membrane of Pseudomonas aeruginosa, a common bacterium that can cause infections in humans. OprI is part of a family of proteins called Opr proteins, which are involved in various cellular processes including nutrient uptake and antibiotic resistance. OprI has been shown to play a role in the virulence of P. aeruginosa by promoting bacterial survival and evasion of the host immune response.
Specifications
oprI | |
Polyclonal | |
Unconjugated | |
oprI | |
Major outer membrane lipoproteinoprI | |
Rabbit | |
Affinity chromatography | |
RUO | |
77220643 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P11221 | |
oprI | |
Recombinant Pseudomonas aeruginosa (strain ATCC 15692/DSM 22644/CIP 104116/JCM 14847/LMG 12228/1C/PRS 101/PAO1) oprI protein (1-83aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction