Learn More
Invitrogen™ OPRK1 Polyclonal Antibody

Description
Positive control: rat brain.
Specifications
Specifications
| Antigen | OPRK1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | OPRK1 |
| Gene Accession No. | P34975, P41145 |
| Gene Alias | kappa opioid receptor; kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; OPRK1; Oprk2; Ork1; R21; Ror-d |
| Gene Symbols | OPRK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.