Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14561
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14561 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14561 Supplier Thermo Scientific Supplier No. PA14561
Add to Cart
Add to Cart

OPRM1 Polyclonal Antibody

OPRM1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OPRM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene OPRM1
Gene Accession No. P33535, P35372, P42866
Gene Alias hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; OPRM; Oprm1; Oprrm1; OTTHUMP00000017463; Ror-b
Gene Symbols OPRM1
Host Species Rabbit
Immunogen Synthetic peptide comprising residues N(386) H Q L E N L E A E T A P L P(400) of human, mouse and rat MOR-1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18390, 25601, 4988
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.