Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPRS1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6336 Supplier Bioss Supplier No. BS5111R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems. As indicated by its previous name, opioid receptor sigma 1 (OPRS1), the product of this gene was erroneously thought to function as an opioid receptor; it is now thought to be a non-opioid receptor. Alternative splicing of this gene results in transcript variants encoding distinct isoforms.

Specifications

Antigen OPRS1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIGMAR1
Gene Accession No. Q99720
Gene Alias AAG8; Aging-associated gene 8 protein; AL024364; ALS16; DSMA2; hSigmaR1; mSigmaR1; opioid receptor, sigma 1; oprs1; RP23 167I12.6; SIG 1R; SIG-1R; sigma 1 receptor variant SR-1D; Sigma 1-type opioid receptor; Sigma non-opioid intracellular receptor 1; sigma1 receptor; Sigma-1 Receptor; sigma1R; Sigma1-receptor; SIGMAR1; SR BP; SR31747 binding protein 1; SR31747-binding protein; SRBP; SR-BP; SR-BP1; zgc:56378
Gene Symbols SIGMAR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OPRS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10280
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.