Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Opsin 1 (Medium Wave) Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB222194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222194 Supplier Novus Biologicals Supplier No. NBP229858AF350
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Opsin 1 (Medium Wave) Polyclonal antibody specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Opsin 1 (Medium Wave)
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2
Host Species Chicken
Immunogen Synthetic peptides from mouse green opsin and zebra finch red opsin.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2652
Target Species Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.