Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Opsin 1 (Medium Wave) Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$181.90 - $610.00
Specifications
Antigen | Opsin 1 (Medium Wave) |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500:1:2000, ELISA 1:100-1:2000 |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Description
Opsin 1 (Medium Wave) Polyclonal specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Opsin 1 (Medium Wave) | |
Western Blot 1:500:1:2000, ELISA 1:100-1:2000 | |
Polyclonal | |
Chicken | |
Vision | |
PBS with 0.02% Sodium Azide | |
GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2 | |
OPN1MW | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
1 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
RUO | |
Rat, Mouse | |
P04000 | |
2652 | |
Synthetic peptides from mouse green opsin and zebra finch red opsin. | |
Affinity Purified | |
Opsin, red and green. The antibody reacts with a protein of ∼40 kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title