Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Opsin 1 (Medium Wave) Antibody, Novus Biologicals™
SDP

Catalog No. NB018005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018005 0.025 mL
NB11074730 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018005 Supplier Novus Biologicals Supplier No. NB110747300.025ML
Only null left
Add to cart
Add to cart

Rabbit Polyclonal Antibody has been used in 9 publications

Opsin 1 (Medium Wave) Polyclonal specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Opsin 1 (Medium Wave)
Applications Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1: 1000 - 1 : 2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen
Formulation Whole antisera with No Preservative
Gene Accession No. O35599
Gene Alias GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2
Gene Symbols OPN1MW
Host Species Rabbit
Immunogen A synthetic peptide as a part of mouse and rat OPSG (green-sensitive opsin, M opsin, Opn1mw, Gcp) conjugated to immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2652
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.