Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Opsin 1 (Medium Wave) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB11074730
Description
Opsin 1 (Medium Wave) Polyclonal specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Opsin 1 (Medium Wave) | |
Polyclonal | |
Unconjugated | |
Lyophilized from whole antisera with No Preservative | |
GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2 | |
Rabbit | |
Unpurified | |
RUO | |
2652 | |
Mouse, Rat | |
Antisera |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
LYOPH | |
Western Blot 1:250-1:500, Immunohistochemistry 1:250-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:250-1:500, Immunohistochemistry-Frozen 1:250-1:500 | |
O35599 | |
OPN1MW | |
A synthetic peptide from mouse Opsin 1 (Medium Wave) conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat. | |
0.1 mL | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction