Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Opsin 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534040
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Dog, Bat, Bovine, Elephant, Panda, Horse, Rabbit (100%) Gorilla, Rat (94%).
Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
Specifications
Opsin 5 | |
Polyclonal | |
Unconjugated | |
OPN5 | |
DJ402H5.1; G protein-coupled receptor 136; Gpr136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; GRP136; Neuropsin; OPN5; opsin 5; opsin-5; Pgr12; TMEM13; Transmembrane protein 13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100055886, 100353027, 100856777, 221391, 353344, 541280 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q6U736, Q6VZZ7 | |
OPN5 | |
Synthetic 17 amino acid peptide from 7th transmembrane domain of human OPN5. | |
50 μg | |
Primary | |
Human, Mouse, Canine, Rabbit, Monkey, Bovine, Horse, Bat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction