Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Opsin 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534041
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534041 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534041 Supplier Invitrogen™ Supplier No. PA534041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Bovine, Elephant, Horse, Rabbit (100%) Gibbon, Mouse (94%) Bat (81%).

Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin 5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene OPN5
Gene Accession No. Q6U736
Gene Alias DJ402H5.1; G protein-coupled receptor 136; Gpr136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; GRP136; Neuropsin; OPN5; opsin 5; opsin-5; Pgr12; TMEM13; Transmembrane protein 13
Gene Symbols OPN5
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human OPN5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100055886, 100353027, 100856777, 221391, 541280
Target Species Human, Canine, Rabbit, Monkey, Bovine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.