Learn More
Invitrogen™ Opsin 5 Polyclonal Antibody

Description
Positive Samples: U-251MG, HeLa, Mouse eye, Mouse brain, Mouse testis, Rat eye, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | Opsin 5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | OPN5 |
| Gene Accession No. | Q6U736, Q6VZZ7 |
| Gene Alias | DJ402H5.1; G protein-coupled receptor 136; Gpr136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; GRP136; Neuropsin; OPN5; opsin 5; opsin-5; Pgr12; TMEM13; Transmembrane protein 13 |
| Gene Symbols | OPN5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.