Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Opsin Blue Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564948 Supplier Invitrogen™ Supplier No. PA564948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RALKAVAAQQQ ESATTQKAER EVSRM Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.

Opsin Blue belongs to the G-protein coupled receptor 1 family, opsin subfamily. It encodes the blue cone pigment gene which is one of three types of cone photoreceptors responsible for normal color vision. Defects in this gene are the cause of tritan color blindness (tritanopia). Affected individuals lack blue and yellow sensory mechanisms while retaining those for red and green. Defective blue vision is characteristic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin Blue
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OPN1SW
Gene Accession No. P03999
Gene Alias AW551857; BCP; blue cone opsin; blue cone opsin-like pigment; blue cone photoreceptor pigment; blue cone pigment; blue opsin; blue/UV opsin; blue-sensitive opsin; BOP; CBT; OPN1SW; opsin 1 (cone pigments), short-wave-sensitive; opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan); s opsin; short wavelength sensitive opsin; short wavelength-sensitive cone opsin; Short-wave-sensitive opsin 1; SWS opsin; UV cone pigment
Gene Symbols OPN1SW
Host Species Rabbit
Immunogen Recombinant Human Opsin Blue. Recombinant protein control fragment (Product #RP-105728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.