Learn More
Invitrogen™ Opsin Blue Polyclonal Antibody
Description
Predicted Reactivity: Human (100%), Chimpanzee (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Opsin Blue |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | OPN1SW |
| Gene Accession No. | P03999, P51491 |
| Gene Alias | AW551857; BCP; blue cone opsin; blue cone opsin-like pigment; blue cone photoreceptor pigment; blue cone pigment; blue opsin; blue/UV opsin; blue-sensitive opsin; BOP; CBT; OPN1SW; opsin 1 (cone pigments), short-wave-sensitive; opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan); s opsin; short wavelength sensitive opsin; short wavelength-sensitive cone opsin; Short-wave-sensitive opsin 1; SWS opsin; UV cone pigment |
| Gene Symbols | OPN1SW |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.