Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Opsin Green Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107203
Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal. May increase spectral sensitivity in dim light. Expresssed in cone photoreceptor cells.
Specifications
Opsin Green | |
Polyclonal | |
Unconjugated | |
OPN1MW | |
CBBM; CBD; COD5; cone dystrophy 5 (X-linked); GCP; GOP; green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3; opsin 1 (cone pigments), medium-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan); opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein | |
Rabbit | |
Affinity chromatography | |
RUO | |
14539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
O35599 | |
OPN1MW | |
Synthetic peptide located within the following region: CYLQVWLAIR AVAKQQKESE STQKAEKEVT RMVVVMVFAY CLCWGPYTFF. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction