Learn More
Invitrogen™ Opsin Green Polyclonal Antibody

Description
This antibody is predicted to react with chicken, rat and Xenopus based on sequence homology.
Specifications
Specifications
| Antigen | Opsin Green |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OPN1MW |
| Gene Accession No. | P04001 |
| Gene Alias | CBBM; CBD; COD5; cone dystrophy 5 (X-linked); GCP; GOP; green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3; opsin 1 (cone pigments), medium-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan); opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein |
| Gene Symbols | OPN1MW |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.