Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Opsin Green Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598031
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598031 Supplier Invitrogen™ Supplier No. PA598031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Opsin Green Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal. May increase spectral sensitivity in dim light. Expresssed in cone photoreceptor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin Green
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OPN1MW
Gene Accession No. P04001
Gene Alias CBBM; CBD; COD5; cone dystrophy 5 (X-linked); GCP; GOP; green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3; opsin 1 (cone pigments), medium-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan); opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein
Gene Symbols OPN1MW
Host Species Rabbit
Immunogen Recombinant Human Medium-wave-sensitive opsin 1 protein (1-52AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2652
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.