Learn More
Gibco™ Opti-Plex™ Complexation Buffer
Description
Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium. It is designed for complexation of ExpiFectamine transfection reagents for both standard transfection applications and transfections to generate methionine-labeled proteins.
Opti-Plex Complexation Buffer features include:
- Contains recombinant insulin, recombinant transferrin, and GlutaMAX Supplement
- Does not contain methionine, biotin, or phenol red
- A part of the ExpiFectamine293 Met (–) Transfection Kit (Cat. No. A39249)
Specifications
Specifications
| Chemical Name or Material | Complexation Buffer |
| For Use With (Application) | Protein Expression |
| Physical Form | Liquid |
| Quantity | 100 mL |
Safety and Handling
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.