Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ OPTI-SOLV™ Mini, Micro, and Nano Filter
The OPTI-SOLV™ Mini Filter provides low-impact filtering in a package no longer that a finger tight fitting. Use them to prolong the life of your analytical column, or before your mass spectrometer as a last line of defense against debris.
$458.00 - $506.00
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
11-100-9085
|
MilliporeSigma™ Supelco™
51175U |
Micro filter, 10 μm, 5/Pk. |
Each for $506.00
|
|
|||||
11-100-9223
|
MilliporeSigma™ Supelco™
51168U |
Mini filter, 0.5 μm, 5/Pk. |
Each for $460.00
|
|
|||||
11-100-9224
|
MilliporeSigma™ Supelco™
51170U |
Mini filter, 2 μm, 5/Pk. |
Each for $458.00
|
|
|||||
Description
The OPTI-SOLV™ Micro Filter is based on the same design as the Mini Filter and cuts the internal volume to less that 200 nL. It retains the Mini Filter′s ease of use and functionality. The OPTI-SOLV™ Micro Filter features a zero-dead volume connection utilizing Optimize Technologies™ patented floating stem technology.Specifications
Liquid Chromatography |
OPTI-SOLV is a registered trademark of Optimize Technologies, Inc.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title