Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Agar Gel Media Transport Tube with Miniature Rayon Tip Swab, Charcoal Additives

Catalog No. p-7111633
Click to view available options
Shaft Material:
Aluminum
Polystyrene
Soft Aluminum
Closure Color:
Black
Orange
White
This item is not returnable. View return policy

Catalog No. 22-027-139

Supplier: Puritan™ ACH61SAR

This item is not returnable. View return policy

For preservation of aerobic, anaerobic, fastidious bacteria

4 ml Amies Gel w/ Charcoal. Aerobic and Anaerobic CultureFor aerobes, anerobes, and fastidious bacteria. Miniature tip helps minimize discomfort. Ear, nose, throat and eye sampling. Male ureathral sampling. Shelf Life: 24 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Closure Color Black
Closure Type Screw Cap
Description Diameter: 5.15mm
Diameter (Metric) 5.15 mm
For Use With (Application) Preservation of samples
Includes Charcoal additives
Shaft Material Soft Aluminum
Tip Material Rayon
Tip Type Standard
Tube Size 154 mm
Volume (Metric) 4 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.