Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ Opti-Tranz™ Liquid Amies Media Transport Tube with Miniature Rayon Tip Swab
Click to view available options
Tip Type:
Dual applicator
Minitip
Single applicator
Closure Color:
Orange
Red
White
Description
1 ml Liquid Amies in absorbent polyurethane foam, which acts as a moisture reservoir for bacteria. Intended for collection and transport of aerobic and fastidious bacteria. Used in bacterial culture, rapid tests, and Gram staining. Soft rayon tips are non-toxic and made from USP grade fiber. Miniature tip causes minimal patient discomfort. Ear, nose, throat, and eye sampling, as well as male urethral sampling. Shelf Life: 18 Months. Made in USA
Specifications
Specifications
Product Type | Transport Tube |
Media Type | Semi-solid (Sodium Thioglycollate and Agar) |
Sterility | Sterile |
Closure Color | Orange |
Closure Type | Screw Cap |
For Use With (Application) | Throat, urogential, and wound swab sampling |
Shaft Material | Aluminum |
Tip Material | Rayon |
Tip Type | Minitip |
Tube Size | 154 mm |
Show More |
For in vitro diagnostic (IVD) use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction