Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Puritan™ Opti- Tranz™ Liquid Amies Media Transport Tube with Miniature Rayon Tip Swab, Aluminum Handle
                    Click to view available options
                
                
                    
                            
                                Fill Volume:
                            
                        
                        
                            
                                1 mL
                            
                        
                    Description
1 mL Liquid Amies in absorbent polyurethane foam.  Aerobic CultureIdeal for direct microscopic examination of clinical samples.  Rayon tips are non-toxic and made from USP grade fiber.  Soft aluminum wire is flexible and can be easily shaped.  Ear, nose, throat, and eye sampling, as well as male urethral sampling.  Shelf Life: 18 Months.   Made in USA
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Product Type | Transport Tube | 
| Media Type | Semi-solid (Sodium Thioglycollate and Agar) | 
| Sterility | Sterile | 
| Closure Color | Green | 
| Closure Type | Screw Cap | 
| Fill Volume | 1 mL | 
| For Use With (Application) | Throat, urogential, and wound swab sampling | 
| Shaft Material | Soft Aluminum | 
| Tip Material | Rayon | 
| Tip Type | Minitip | 
| Show More | 
For in vitro diagnostic (IVD) use only.
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction