Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Liquid Stuart Media Transport Tube with HydraFlock™ Tip Swab

Features three-dimensional microstructure fiber design for sampling

Specifications

Tube Size 154mm
Volume (Metric) 1mL
Handle Material Polystyrene
Media Type Semi -solid (Sodium thioglycollate and agar)
Product Type Transport tube
View More Specs

Products 2
Catalog Number Mfr. No. Tip Type Closure Color Price Quantity & Availability  
Catalog Number Mfr. No. Tip Type Closure Color Price Quantity & Availability  
22-027-130 View Documents Puritan™
LS61H
Single Flock Yellow This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-027-131 View Documents Puritan™
LS62H
Double flock Pink This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Maintains viability of microorganisms without significant increase in growth
  • Non-nutritive general purpose medium
  • Sodium thioglycollate as reducing agent, sodium glycerophosphate to buffer calcium chloride
  • 1mL sterile liquid stuart gel medium
  • Highly absorbent soft smooth tip (Length: 19.56mm, Dia.: 5.56mm)
  • Handle Length: 149.61mm, Dia.: 2.54mm

Applications

  • Aerobic culture
  • Ear, nose, throat, and eye sampling, as well as male urethral sampling
  • Ideal for direct microscopic examination of clinical samples
Specifications

Specifications

154mm
Polystyrene
Transport tube
Screw
1mL
Semi -solid (Sodium thioglycollate and agar)
Throat, urogential, and wound swab sampling
Safety and Handling

Safety and Handling

ShelfLife : 18 months

For in vitro diagnostic (IVD) use only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit