Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Opticin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537262
Description
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
Opticin, also known as OPTC or OPT, is a 332 amino acid protein that is secreted into the extracellular matrix and contains seven LRR repeats. Expressed in skin and fetal liver and also present in ocular tissues, such as the retina, cornea and iris, opticin functions to bind collagen fibrils and, via this interaction, is thought to regulate fibril spacing, morphology and organization. Opticin is subject to post-translational O-glycosylation and may be associated with the pathogenesis of primary open angle glaucoma.
Specifications
| Opticin | |
| Polyclonal | |
| Unconjugated | |
| OPTC | |
| dermatan sulfate proteoglycan 3; dspg3; Oculoglycan; OPT; Optc; optcl; opticin; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26254, 269120, 304802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q920A0, Q9UBM4 | |
| OPTC | |
| A synthetic peptide corresponding to residues in human Opticin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction