Learn More
Invitrogen™ Opticin Polyclonal Antibody

Description
Immunogen sequence: ASLPRKERKR REEQMPREGD SFEVLPLRND VLNPDNYGEV IDLSNYEELT DYGDQLPEVK VTSLAPATSI SPAKSTTAPG TPSSNPTMTR PTTAGL.
Specifications
Specifications
| Antigen | Opticin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | OPTC |
| Gene Accession No. | Q920A0 |
| Gene Alias | dermatan sulfate proteoglycan 3; dspg3; Oculoglycan; OPT; Optc; optcl; opticin; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 |
| Gene Symbols | OPTC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.