Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Opticin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111046 Supplier Invitrogen™ Supplier No. PA5111046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPRKERKRRE EQMPREGDSF EVLPLRNDVL NPDNYGEVID LSNYEELTDY GDQLPEVKVT SLAPATSISP Highest antigen sequence indentity to the following orthologs: Mouse - 50%, Rat - 46%.

Opticin, also known as OPTC or OPT, is a 332 amino acid protein that is secreted into the extracellular matrix and contains seven LRR repeats. Expressed in skin and fetal liver and also present in ocular tissues, such as the retina, cornea and iris, opticin functions to bind collagen fibrils and, via this interaction, is thought to regulate fibril spacing, morphology and organization. Opticin is subject to post-translational O-glycosylation and may be associated with the pathogenesis of primary open angle glaucoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opticin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OPTC
Gene Accession No. Q9UBM4
Gene Alias dermatan sulfate proteoglycan 3; dspg3; Oculoglycan; OPT; Optc; optcl; opticin; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047
Gene Symbols OPTC
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Opticin. Recombinant protein control fragment (Product #RP-104649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26254
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.