Learn More
Description
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.
- Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization
- Surface area per gram of a particle varies inversely with the particle diameter
- Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based
- For most applications, particles can be used directly from the bottle without any pre-cleaning
- Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces or cause proteins to desorb from particle surfaces
- Various surface acid concentrations available to help optimize reagent development efforts
- Particles have been used in and are equally successful in molecular biology and research applications
Parking Area Properties
- Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
- Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
- High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling
Recommended for:
- Clinical diagnostics
- Molecular biology
- Nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
Specifications
Specifications
| Additive | Proprietary anionic surfactant |
| Certifications/Compliance | Includes Certificate of Analysis |
| Concentration | 4% |
| Content And Storage | Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
| Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
| Color | White or Colorless |
| For Use With (Application) | Covalent Coupling |
| Material | Polystyrene Sulfate |
| Product Type | Specialty Particles |
| Includes | Package Insert Sheet containing Certicate of Analysis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.