Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Optimized Fermentation of Yeast Kit
GREENER_CHOICESCI_ED

Catalog No. s37747 Shop All Aldon Products
Click to view available options
Class Size:
15 Lab Groups

Includes

50g Glucose, 50g Sucrose, 50g Lactose, 50g Fructose, 10g Yeast, Teacher's Guide and Student copymasters

Students study effectiveness of fermentation process on different types of sugars by monitoring growth rate and respiration to determine the optimum food source for yeast.

Yeast will be exposed to four different sugar solutions and the overall pressure changes for fermentation in each sugar solution will be compared.

  • Contains enough materials for 15 groups of students

Specifications

Product Type Optimized Fermentation of Yeast Kit
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To study effectiveness of fermentation process on different types of sugars by monitoring growth rate and respiration to determine the optimum food source for yeast.
Includes 50g Glucose, 50g Sucrose, 50g Lactose, 50g Fructose, 10g Yeast, Teacher's Guide and Student copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.