Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Optineurin Antibody (08), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306529AF700
Description
Optineurin Monoclonal antibody specifically detects Optineurin in Human samples. It is validated for ELISASpecifications
Optineurin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
08 | |
ELISA | |
FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Optineurin (NP_068815.2; Met1-Ile577). | |
0.1 mL | |
Immunology, Innate Immunity, Vision | |
10133 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction